Caps for sample tubes, tamper evident
Screw cap, with lip seal for tamper evident tubes, 30 ml, blanc
N° de catalogue SIMPT552-30WTP
Fournisseur: Simport Scientific
Caps for sample tubes, tamper evident
N° de catalogue SIMPT552-30WTP
Fournisseur: Simport Scientific
Aller à :
Product Details & Documents
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Spécifications
- Pour tubes:30 ml
- Couleur:blanc
- Désignation:Screw cap, with lip seal for tamper evident tubes
- Seq:06