Chromatography data station, DataApex ClarityVA
Chromatography data station, DataApex ClarityVA
Clarity Chromatography Data Station Lite version for analogue connection of one chromatography instrument. Data Processing only, no instrument control is included
N° de catalogue 906-0162
Fournisseur:   COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
N° de catalogue 906-0162
Fournisseur:  COLLECTION PREFIX HPLC
Chromatography data station, DataApex ClarityVA
Chromatography data station, DataApex ClarityVA
Clarity Chromatography Data Station Lite version for analogue connection of one chromatography instrument. Data Processing only, no instrument control is included
Prix
1 610,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
ClarityVA is a flexible and powerful Chromatography Data System (CDS) that controls HPLC and GC systems. This modular software includes a lot of controls for many different systems, modules and interfaces. The flexibility enables an easy control of HPLC systems in combination with a user friendly surface.
  • Direct control of Hitachi HPLC (Chromaster and Primaide)
  • Modules for specific calculation and methodology
  • Tools for regulated environment
  • Extended user support
  • Language localisations (English, German, French,...)
ClarityVA is useable for modern complained pharma labs, when configured correctly (21 CFR part 11 compliance).
Documents
Spécifications
  • Désignation:
    Clarity Chromatography Data Station Lite version for analogue connection of one chromatography instrument. Data Processing only, no instrument control is included
  • Seq:
    15
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.