Chromatography columns with filter plates
Chromatography column with PTFE tap, 60,00 mm, 600 mm, 45/40 NS
N° de catalogue NEUB1202450600600
Fournisseur: NEUBERT VOLUME GLASSWAERE
Chromatography columns with filter plates
N° de catalogue NEUB1202450600600
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
These chromatography columns feature a top sleeve with PTFE tap and are made from borosilicate glass 3.3.
Spécifications
- Ø int.:60,00 mm
- Rodage femelle:45/40 NS
- Désignation:Chromatography column with PTFE tap
- Longueur:600 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



