Electron multipliers
Electron multiplier for 7700 ICP-MS
N° de catalogue HEWL5190-0108
Fournisseur: Agilent
Aller à :
Product Details & Documents
The electron multiplier converts incident ions into an amplified electric current used to record measurements of the ion signal in ICP-MS Systems.
As the voltage in the system increases the amplification factor (or gain) increases. In general the electron multiplier can measure a count of 1.6E10. The system should be replaced when the total ion count reaches more than 3.2E11 and the sensitivity is less than 4000 counts with 10 ppb Y (even when applying detector voltage above 2700 V).
Spécifications
- Désignation:Electron multiplier for 7700 ICP-MS
- Seq:01