Safety caps
Safety cap II (includes 1 PP blue basic cap with 2 ports and 2 shut-off valves, GL 45, with PTFE cone, 2 PFA fittings, 3,2 mm (1 blue, 1 red), 2 white PTFE fittings, 3,2 mm, 1 venting valve, including air filter, 1 μm, PTFE membrane), for GL 45 threaded bottles
HEWL5043-0224
: Agilent
These safety caps have been designed for optimal sealing with an integrated exhaust valve providing pressurisation during solvent extraction and allowing proper solvent flow to your HPLC system.
- Caps rotate freely, preventing tube twisting during bottle exchange
- Constructed of PTFE and PFA for high chemical resistance
The exhaust valve contains a PTFE membrane to prevent contamination of your solvents from dirt and dust particles.
- Désignation:Safety cap II (includes 1 PP blue basic cap with 2 ports and 2 shut-off valves, GL 45, with PTFE cone, 2 PFA fittings, 3,2 mm (1 blue, 1 red), 2 white PTFE fittings, 3,2 mm, 1 venting valve, including air filter, 1 μm, PTFE membrane), for GL 45 threaded bottles
- Seq:01