Capillary columns, HP-PLOT MoleSieve
Columns, HP-PLOT MoleSieve, Capillaire, HP-PLOT Molesieve, 25,00 µm, 0,32 mm, 30 m
N° de catalogue HEWL19091P-MS8LTM
Fournisseur: Agilent
Aller à :
Product Details & Documents
For applications requiring resolution of argon and oxygen without cryogenic cooling, select columns with thick film. The thin film configurations are ideal for many applications, including routine air monitoring and analysis. The thin film column resolves argon and oxygen at subambient temperatures.
Spécifications
- Type de colonne:Capillaire
- Épaisseur du film:25,00 µm
- Ø int.:0,32 mm
- Désignation:Columns, HP-PLOT MoleSieve
- Longueur:30 m
- Seq:05
- Greffage:HP-PLOT Molesieve