M Stand for Bio-bin® Boxes
M Stand for Bio-bin® Boxes
M stand for 30 l Bio-bin® boxes, stainless steel, for floor use
N° de catalogue ECONEXBBMS30.0
Fournisseur:   ECONIX LTD
M Stand for Bio-bin® Boxes
N° de catalogue ECONEXBBMS30.0
Fournisseur:  ECONIX LTD
M Stand for Bio-bin® Boxes
M Stand for Bio-bin® Boxes
M stand for 30 l Bio-bin® boxes, stainless steel, for floor use
Prix
83,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These stainless steel stands were designed to allow the 2, 5 and 30 Litre Bio-bin® boxes to tilt at approximately 15 degrees to allow easy filling for tall thin items e.g. pipettes.
  • Allow the user to see the contents easily while helping to prevent overfilling and also 'wigwamming' of items
  • Small stand for 2 and 5 L boxes is ideal for use on work benches
  • Larger 30 L M stand is perfect for floor use
Spécifications
  • Désignation:
    M stand for 30 l Bio-bin® boxes, stainless steel, for floor use