Pouring rings
Pouring ring, GL 80, incolore
Catalog # BIBB4516/80D
Supplier: SciLabware
Jump to:
Product Details & Documents
Pouring rings, made of Polypropylene, for laboratory/media bottles.
- Ensures drip-free pouring
- Very good chemical resistance
- Autoclavable up to 140 °C
Specifications
- Filetage:GL 80
- Couleur:incolore
- Désignation:Pouring ring
- Seq:03