Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Spray gun

Spray gun

Replacement cap, orange
N° de catalogue ENTE600037OR
Fournisseur:   ENTEGRIS
Spray gun
N° de catalogue ENTE600037OR
Fournisseur:  ENTEGRIS
Spray gun
Spray gun

Replacement cap, orange

Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

High purity PFA and PTFE (all wetted parts).

  • Orifice: 4,8 mm
  • Connection: 1/4" or 1/2" FNPT
  • Pressure: max. 5516 mbar

Used in fume hoods and wet processing stations to help protect against corrosion.

Spécifications
  • Désignation:
    Replacement cap, orange
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.