Buses de pulvérisation
Nozzle for spray gun
N° de catalogue ENTE215-28
Fournisseur: ENTEGRIS
Aller à :
Product Details & Documents
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
Spécifications
- Désignation:Nozzle for spray gun