Grounding cables, Super Erecta®
Grounding cable with universal bracket
N° de catalogue METOGCB16SA
Fournisseur: Metro International
Aller à :
Product Details & Documents
These grounding cables provide an effective method of electrostatic discharge for any Metro Super Erecta unit (stationary or mobile) when used in conjunction with aluminum split sleeves or ESD kit and conductive casters.
- Cables ensure a proper path to ground when used with ESD flooring
- Grounding cable with universal bracket fits wire and solid shelves, as well as open-wire frames
Spécifications
- Désignation:Grounding cable with universal bracket
- Seq:02