Multiple wafer shippers
Assembly, cleaned, Wafershield™ high purity, Wafer thickness 150 - 675 µm, PP
N° de catalogue ENTEH9150-0038
Fournisseur: ENTEGRIS
Aller à :
Product Details & Documents
Ultrapak® series multiple wafer shipping boxes are made of Ultrapure polypropylene materials that provide a clean shipping environment and assure low levels of condensable inorganics.
- Horizontal and vertical robotic pickup flanges on the cassette allow convenient handling
- Center notch track alignment and 'H' bar ensure accurate equipment interoperability both vertically and horizontally
- Individual horizontal cantilever springs limit wafer rotation to reduce particle contamination and also secure the wafers in the cassette to safeguard against breakage
- Three design options to accommodate varying wafer thickness
Spécifications
- Pour:Wafer thickness 150 - 675 µm
- Désignation:Assembly, cleaned, Wafershield™ high purity
- Matière:PP