HPLC columns, Acclaim™ Trinity™ P2
Cartouche de garde, 3 µm, 3,00 mm, 10 mm
N° de catalogue 555-7337
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.
- Desired selectivity for pharmaceutical counterion screening
- Retention of ionic and ionisable analytes without using ion-pairing reagents
- Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods
Particle type: ultra pure, spherical silica
Max. pressure: 6000 psi
Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats
Bonded phase: anion-exchange, cation exchange and HILIC
pH range: 2,0 to 8,0
Spécifications
- Type de colonne:Cartouche de garde
- Ø int.:3,00 mm
- Granulométrie:3 µm
- Longueur:10 mm
- Seq:06
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- 554-4528Porte-cartouche de garde en acier inoxydable, V-2Connexion Pour la disponibilité1 * 1 ST - 415,00 €



