Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, left hinged, 600×366×780 mm
N° de catalogue KOTT307-00060
Fournisseur:   KOETTERMANN
Wall-mounted cabinets
N° de catalogue KOTT307-00060
Fournisseur:  KOETTERMANN
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, left hinged, 600×366×780 mm

Prix
422,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Spécifications
  • Désignation:
    Wall-mounted cabinet, with one shelf
  • Seq:
    30
  • l×p×h:
    600×366×780 mm
  • Portes:
    1 door, left hinged