Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
DNA polymerases, Phusion® U hot start

DNA polymerases, Phusion® U hot start

Phusion® U hot start DNA polymerase, 100 units, 1 KIT
N° de catalogue F-555S
Fournisseur:   FINNZYMES REAGENTS
DNA polymerases, Phusion® U hot start
N° de catalogue F-555S
Fournisseur:  FINNZYMES REAGENTS
Numéro CAS:  
DNA polymerases, Phusion® U hot start
DNA polymerases, Phusion® U hot start

Phusion® U hot start DNA polymerase, 100 units, 1 KIT

Numéro CAS:  
Prix
212,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

Phusion® U DNA polymerase is a novel engineered high fidelity enzyme developed using fusion technology.

  • Accuracy: high fidelity DNA polymerase (25X Taq)
  • Uracil-tolerance: engineered to incorporate dUTP and amplify uracil containing templates
  • Specificity: hot start for reduced nonspecific amplification and primer degradation
  • Speed: short extension times (15 to 30 s/kb)
  • Store at –20 °C

Due to a proprietary mutation in the so called dUTP binding pocket of Phusion®, Phusion® U overcomes an important limitation of proofreading enzymes - it can incorporate dUTP and read through uracil present in DNA templates.

In addition to uracil usage possibility, it features all the superior properties of other Phusion® DNA polymerases - great accuracy, speed, ability to amplify long amplicons up to 20 kb and a high specificity with Affibody based hot start. These features make DNA Polymerase an excellent choice for such important applications as amplification of bisulphite converted or damaged DNA as well as use of carryover contamination control.

It is also available in the master mix and green formats. Phusion® U hot start PCR master mix (FINRF-533S and FINRF-533L) is convenient 2X mix designed to minimise the number of pipetting steps. Only template and primers need to be added by the user. Phusion® U green format (FINRF-556S and FINRF-556L) is a combination of Phusion® U hot start DNA polymerase and 5X Phusion® green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The coloured buffer does not interfere with Phusion® U performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Delivery information: Includes: Phusion® U hot start DNA polymerase, 2 Units/µl; 5X Phusion® HF buffer; 5X Phusion® GC buffer; DMSO. Both Phusion® HF buffer and Phusion® GC buffer provide 1,5 mm MgCl₂ in the final 1X concentration.

Caution: For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Cdt:
    1 KIT
  • Désignation:
    Phusion® U hot start DNA polymerase, 100 units

Spécifications