Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Handheld microscope with exchangeable caps + polariser, Edge, UNIVERSAL, AM4815ZT, 20 - 220×, 8 LEDs, 1,3 MP, EDOF/EDR
N° de catalogue 620-2622
Fournisseur:   Dino-Lite / IDCP B.V.
Handheld Digital Microscopes, Universal
N° de catalogue 620-2622
Fournisseur:  Dino-Lite / IDCP B.V.
Handheld Digital Microscopes, Universal
Handheld Digital Microscopes, Universal
Handheld microscope with exchangeable caps + polariser, Edge, UNIVERSAL, AM4815ZT, 20 - 220×, 8 LEDs, 1,3 MP, EDOF/EDR
Prix
912,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
  • High speed USB 2.0 connection
  • Metal or composite housing
  • Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Livraison: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : [email protected]
Spécifications
  • Grossissement:
    20 - 220×
  • Modèle:
    UNIVERSAL, AM4815ZT
  • Résolution:
    1,3 MP
  • Désignation:
    Handheld microscope with exchangeable caps + polariser, Edge
  • Seq:
    15
  • Caractéristiques supplémentaires:
    EDOF/EDR
  • Éclairage:
    8 LEDs
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • IDCPRK-06-PL
    Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)
    Connexion Pour la disponibilité
    1 * 1 ST - 28,60 €
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • 630-1821
    Foot pedal, enables users to take a picture with their foot
    Connexion Pour la disponibilité
    1 * 1 ST - 63,30 €
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • 630-2517
    Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    Connexion Pour la disponibilité
    1 * 1 ST - 340,00 €
Informations sur le produit
  • N° de catalogue
    Désignation
    l×p×h
    Disponibilité
    Prix
  • IDCPRK-06A
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    150×220×270 mm
    Connexion Pour la disponibilité
    1 * 1 ST - 252,00 €
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • 630-2923
    Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery
    Connexion Pour la disponibilité
    1 * 1 ST - 329,00 €
Informations sur le produit
  • N° de catalogue
    Désignation
    l×p×h
    Disponibilité
    Prix
  • IDCPRK-06AE
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EX
    150×220×270 mm
    Connexion Pour la disponibilité
    1 * 1 ST - 249,00 €