Scissors No. 145, Feather®
Sharp/sharp
  FEAT208000157
Scissors No. 145, Feather®
  FEAT208000157
Scissors No. 145, Feather®
Scissors No. 145, Feather®
Sharp/sharp
59,90 €
1 * 5 PAIRE
Pathology scissors made of stainless steel with easily exchangeable blades.
  • No regrinding
  • Easy blade change
  • Type:
    Sharp/sharp