Connection system GL 45
Connection system GL 45
Membrane screwcap, PP, blue, GL 45, with 0,2 μm PTFE membrane
N° de catalogue BIBB4507/45D
Fournisseur:   SciLabware
Connection system GL 45
N° de catalogue BIBB4507/45D
Fournisseur:  SciLabware
Connection system GL 45
Connection system GL 45
Membrane screwcap, PP, blue, GL 45, with 0,2 μm PTFE membrane
Prix
42,90 €
Connexion Pour la disponibilité
1 * 5 ST
Product Details & Documents
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
  • Temperature resistant to 140 °C
  • Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
  • Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membrane filter with 0,2 μm sized pores permits sterile pressure equalisation. Unused ports can be sealed with a blanking cap. Components are fully interchangeable and compatible with both two and three port screwcaps.
Spécifications
  • Désignation:
    Membrane screwcap, PP, blue, GL 45, with 0,2 μm PTFE membrane
  • Seq:
    13