Mandrels for cylindrical bend testers
Mandrels for cylindrical bend testers
Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
N° de catalogue SIMESP1833
Fournisseur:   Industrial Physics
Mandrels for cylindrical bend testers
N° de catalogue SIMESP1833
Fournisseur:  Industrial Physics
Mandrels for cylindrical bend testers
Mandrels for cylindrical bend testers
Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
Prix
549,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Documents
Spécifications
  • Désignation:
    Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
  • Seq:
    907