Tissue grinders, CS1, KIMBLE®
50 ml
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
- Narrow pestles eliminate the potential of sample overflow
- Convenient and easy-to-use
- Gamma sterilised in an easy-open Tyvek® pack
The molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination. Tubes are made of PP, pestles are 30% glass-filled PP and caps are HDPE.
Livraison: Each tissue grinder is supplied with a fully assembled pestle, a conical sample tube with solid top cap, an adhesive identification label and an instruction card.
Documents
Spécifications
- Capacité:50 ml
- Seq:02