Inoculating loops, Rotilabo®
Inoculating loop holder set, stainless steel, for inoculating loops with wire of Ø 0,6 - 1,0 mm, assortiment, 245 mm
N° de catalogue ROTHKL97.1
Marketsource
Fournisseur: Roth Carl
Aller à :
Product Details & Documents
Rotilabo® inoculating loops, made of platinum/iridium. Wire diameter 0,6 mm. Rotilabo® inoculating loop holder set made of stainless steel. Loops are fixed with union nut. For inoculating loops with wire diameter 0,6 to 1,0 mm. Set sorted by colour: blue, yellow, red, black. Length: 245 mm. Autoclavable at 121 °C.
Livraison: Loops delivered without inoculating loop holder, please order separately.
Spécifications
- Couleur:assortiment
- Désignation:Inoculating loop holder set, stainless steel, for inoculating loops with wire of Ø 0,6 - 1,0 mm
- Longueur:245 mm
- Seq:02