Mops, PowerHead™
PowerHead™ NovaPoly™ covered foam, 254 mm
N° de catalogue MICRPH6-150
Fournisseur: MICRONOVA
Aller à :
Product Details & Documents
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
- Stainless steel PowerHead™ handles hold and wring the various mop heads
- Autoclavable
- A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Informations de commande: All components are ordered separately.
Spécifications
- Désignation:PowerHead™ NovaPoly™ covered foam, 254 mm
- Seq:11
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.