Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Test tubes with screw caps

Test tubes with screw caps

15 ml, 120 mm, 16 mm, Individually packed, Stérile
N° de catalogue KART88207
Fournisseur:   KARTELL
Test tubes with screw caps
N° de catalogue KART88207
Fournisseur:  KARTELL
Test tubes with screw caps
Test tubes with screw caps

15 ml, 120 mm, 16 mm, Individually packed, Stérile

Prix
495,00 €
Connexion Pour la disponibilité
1 * 700 ST
Product Details & Documents

PS, clear, wtih screw cap.

  • Tight screw cap
  • Good chemical resistance
  • Good temperature resistance

Taking into consideration the different applications, they are available sterile single packed, sterile nultiple packed and not sterile.

Spécifications
  • Ø ext.:
    16 mm
  • Emballage:
    Individually packed
  • Stérilité:
    Stérile
  • Capacité:
    15 ml
  • Longueur:
    120 mm