Separating Funnels, Squibb
Separating Funnels, Squibb
1000 ml, 29/32 NS
N° de catalogue KAVA632426355946
Fournisseur:   Kavalier
Separating Funnels, Squibb
N° de catalogue KAVA632426355946
Fournisseur:  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
1000 ml, 29/32 NS
Prix
700,00 €
Connexion Pour la disponibilité
1 * 2 ST
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Spécifications
  • Rodage femelle:
    29/32 NS
  • Capacité:
    1000 ml
  • Seq:
    05