Separating Funnels, Squibb
250 ml, 29/32 NS
Catalog # KAVA632426355256
Supplier:   Kavalier
Separating Funnels, Squibb
Catalog # KAVA632426355256
Supplier:  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
250 ml, 29/32 NS
Price
299,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Specifications
  • Rodage femelle:
    29/32 NS
  • Capacité:
    250 ml
  • Seq:
    03