Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Lead Standard Solutions, NIST

Lead Standard Solutions, NIST

Plomb, Pb, 1000 mg/L, 100 mL
N° de catalogue HACH1279642
Fournisseur:   Hach
Lead Standard Solutions, NIST
N° de catalogue HACH1279642
Fournisseur:  Hach
Numéro CAS:  
Lead Standard Solutions, NIST
Lead Standard Solutions, NIST

Plomb, Pb, 1000 mg/L, 100 mL

Numéro CAS:  
Prix
45,10 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents

Standard solutions for accuracy checks in lead determinations.

Spécifications

Spécifications

  • Concentration:
    1000 mg/L
  • Cdt:
    100 mL
  • Élement:
    Plomb
  • Symbole:
    Pb

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.