Nessler cylinders
Nessler cylinders
Nessler cylinder 50 ml, low form, with optically plane bottom, circular mark at 50 ml, Ø×L: 26×150 mm
N° de catalogue WITG2.151.050
Fournisseur:   witeg Labortechnik
Nessler cylinders
N° de catalogue WITG2.151.050
Fournisseur:  witeg Labortechnik
Nessler cylinders
Nessler cylinders
Nessler cylinder 50 ml, low form, with optically plane bottom, circular mark at 50 ml, Ø×L: 26×150 mm
Prix
27,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Spécifications
  • Désignation:
    Nessler cylinder 50 ml, low form, with optically plane bottom, circular mark at 50 ml, Ø×L: 26×150 mm
  • Seq:
    04