Accessories for FlashGel™ system
FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
N° de catalogue LONZ57064
Fournisseur:   LONZA
Accessories for FlashGel™ system
N° de catalogue LONZ57064
Fournisseur:  LONZA
Accessories for FlashGel™ system
Accessories for FlashGel™ system
FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
Prix
676,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Spécifications
  • Désignation:
    FlashGel™ recovery kit includes recovery cassettes 1,2%, 8+1 double-tier well, loading dye, recovery buffer, QuantLadder, visualisation glasses, and mask
  • Seq:
    22