Accessories for glove boxes, Precise™
Diaphragm pump for drying train circulation, for 230 V units, 35 l/minute displacement capacity
N° de catalogue LBCP5248201
Fournisseur: LABCONCO
Aller à :
Product Details & Documents
Spécifications
- Désignation:Diaphragm pump for drying train circulation, for 230 V units, 35 l/minute displacement capacity
- Seq:27