Alkaline phosphatase buffer-1 (5X)
Alkaline phosphatase buffer (5X), 100 mL
Fournisseur: Thermo Fisher Scientific
Alkaline phosphatase buffer-1 (5X)
N° de catalogue J62907.AE
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à :
Product Details & Documents
Physical Form: Liquid
Appearance: Clear liquid
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Cdt:100 mL
- Désignation:Alkaline phosphatase buffer (5X)
- Cat. No.:J62907.AE
- Apparence:Clear liquid
- Physical form:Liquid
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



