Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Miconazole nitrate ≥98%

25 g
Catalog # J60796.14
Irritant
Miconazole nitrate ≥98%
Catalog # J60796.14
CAS Number:  22832-87-7
Miconazole nitrate ≥98%
Miconazole nitrate ≥98%

25 g

CAS Number:  22832-87-7
Price
434,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 22832-87-7 Molecular Formula: C18H14Cl4N2O.HNO3 Formula Weight: 479.15 Physical Form: Powder Appearance: White to off-white Melting Point: 170-185° Solubility: Slightly soluble in water. Soluble in propylene glycol or pyridine. MDL No.: MFCD00058161

Applications: Inhibits cytochrome P450-dependent 14a-demethylase

Synonyms:  1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate
  • CAS Number:22832-87-7
  • Formula:C₁₈H₁₄Cl₄N₂O·HNO₃
  • MDL Number:MFCD00058161
  • Molecular Weight:479.14 g/mol
  • Merck Index:14,6178
  • Melting Pt:170…185 °C
  • Storage Temperature:Température ambiante
  • EINECS:245-256-6
Specifications

Specifications

  • Cdt:
    25 g

Specifications