Miconazole nitrate ≥98%
5 g
Fournisseur: Thermo Fisher Scientific
Miconazole nitrate ≥98%
N° de catalogue J60796.06
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 22832-87-7
Aller à :
Product Details & Documents
Purity: 98+% CAS No.: 22832-87-7 Molecular Formula: C18H14Cl4N2O.HNO3 Formula Weight: 479.15 Physical Form: Powder Appearance: White to off-white Melting Point: 170-185° Solubility: Slightly soluble in water. Soluble in propylene glycol or pyridine. MDL No.: MFCD00058161
Applications: Inhibits cytochrome P450-dependent 14a-demethylase
Synonymes: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate
- Numéro CAS:22832-87-7
- Formule:C₁₈H₁₄Cl₄N₂O·HNO₃
- Numéro MDL:MFCD00058161
- Poids moléculaire:479.14 g/mol
- Index Merck:14,6178
- Point de fusion:170…185 °C
- Température de stockage:Température ambiante
- EINECS:245-256-6
Documents
Spécifications
Spécifications
- Cdt:5 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



