Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
100 ml, 24/29 NS
N° de catalogue NEUB0752-15-0100
Flasks, flat bottom with standard ground joint
N° de catalogue NEUB0752-15-0100
Flasks, flat bottom with standard ground joint
100 ml, 24/29 NS
Prix
42,60 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Spécifications
  • Rodage femelle:
    24/29 NS
  • Capacité:
    100 ml
  • Seq:
    02