Flasks, flat bottom with standard ground joint
50 ml, 24/29 NS
N° de catalogue NEUB0752-15-0050
Fournisseur: NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
N° de catalogue NEUB0752-15-0050
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Borosilicate glass 3,3, amber.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Spécifications
- Rodage femelle:24/29 NS
- Capacité:50 ml
- Seq:01