BD Emerald™ Syringes
Seringue sans aiguille, Luer simple, cône centré, 2 ml, Stérile
Fournisseur: BD Medical
Aller à :
Product Details & Documents
Sterile, disposable, three-piece syringes, PP barrel and piston, latex-free stopper. Luer slip syringes available with or without needle. Combining high-quality performance and reduced environmental impact.
- For general purpose use. Specialised design features benefit clinicians, patients and the environment
- Green stopper and bold scale markings to aid in setting an accurate dose
- Clear barrel allows a clear view of fluid
- Retaining ring to help prevent accidental plunger rod pullout
- Flange size and shape to provide stability and comfort during use
- Textured thumb press to reduce slippage during administration
- Minimises waste disposal - has up to 30% less material than other syringes
- Sterile, single use
- Natural rubber latex is not part of the material formulation
Attention: BD Emerald™ 3-piece syringes are not designed for use on syringe pumps.
Spécifications
- Stérilité:Stérile
- Capacité:2 ml
- Désignation:Seringue sans aiguille, Luer simple, cône centré
- Seq:02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



