Washproof plasters, HypaPlast
Washproof plasters, HypaPlast
20 individually wrapped plasters, assorted (38×38 mm and 72×25 mm)
N° de catalogue SAFID7775
Fournisseur:   SAFETY FIRST AID GROUP
Washproof plasters, HypaPlast
N° de catalogue SAFID7775
Fournisseur:  SAFETY FIRST AID GROUP
Washproof plasters, HypaPlast
Washproof plasters, HypaPlast
20 individually wrapped plasters, assorted (38×38 mm and 72×25 mm)
Prix
7,30 €
Connexion Pour la disponibilité
1 * 20 ST
Product Details & Documents
These plasters are ideal for the treatment of wounds, cuts and other minor injuries that do not require a bandage.
  • High quality hypoallergenic plasters
  • Good sticking quality, wash proof
  • Individually wrapped and sterile
  • Available in an assortment of shapes and sizes
Spécifications
  • Désignation:
    20 individually wrapped plasters, assorted (38×38 mm and 72×25 mm)
  • Seq:
    01