Capillary blood collection system
Blood collection system, prepared for serum collection, Coagulation inducer, 39 mm, 11/8 mm, 200 µl
N° de catalogue KABE077101
Marketsource
Fournisseur: KABE LABORTECHNIK
Capillary blood collection system
N° de catalogue KABE077101
Fournisseur: KABE LABORTECHNIK
Marketsource
Aller à :
Product Details & Documents
The capillary blood collection system consists of a prepared test vessel and a prepared plastic end-to-end collection capillary with stopper.
- Easy handling
- Plastic capillary* with exact filling volume; complete inner surface prepared, unbreakable
- Collection vessel serves as centrifugal vessel; prepared for all common tests
Attention: *with conformity certificate in accordance with the Weights and Measurement Regulations
Spécifications
- Ø:11/8 mm
- Volume:200 µl
- Désignation:Blood collection system, prepared for serum collection
- Longueur:39 mm
- Seq:02
- Additive:Coagulation inducer