Capillary blood collection system
Capillary blood collection system
Blood collection system, prepared for serum collection, Coagulation inducer, 39 mm, 11/8 mm, 200 µl
N° de catalogue KABE077101
Marketsource
Fournisseur:   KABE LABORTECHNIK
Capillary blood collection system
N° de catalogue KABE077101
Fournisseur:  KABE LABORTECHNIK
Marketsource
Capillary blood collection system
Capillary blood collection system
Blood collection system, prepared for serum collection, Coagulation inducer, 39 mm, 11/8 mm, 200 µl
Prix
202,20 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
The capillary blood collection system consists of a prepared test vessel and a prepared plastic end-to-end collection capillary with stopper.
  • Easy handling
  • Plastic capillary* with exact filling volume; complete inner surface prepared, unbreakable
  • Collection vessel serves as centrifugal vessel; prepared for all common tests
Attention: *with conformity certificate in accordance with the Weights and Measurement Regulations
Spécifications
  • Ø:
    11/8 mm
  • Volume:
    200 µl
  • Désignation:
    Blood collection system, prepared for serum collection
  • Longueur:
    39 mm
  • Seq:
    02
  • Additive:
    Coagulation inducer