Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
600 ml, 19/26
N° de catalogue NEUB0293-04-0601
Volumetric flasks, class A, blue markings
N° de catalogue NEUB0293-04-0601
Volumetric flasks, class A, blue markings
600 ml, 19/26
Prix
114,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass, grade 3.3, with PE stopper.
  • With one mark
  • Good chemical resistance
  • Good temperature resistance
Spécifications
  • RN:
    19/26
  • Capacité:
    600 ml
  • Seq:
    06