Volumetric flasks, class A, blue markings
600 ml, 19/26
N° de catalogue NEUB0293-04-0601
Fournisseur: NEUBERT VOLUME GLASSWAERE
Volumetric flasks, class A, blue markings
N° de catalogue NEUB0293-04-0601
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Borosilicate glass, grade 3.3, with PE stopper.
- With one mark
- Good chemical resistance
- Good temperature resistance
Spécifications
- RN:19/26
- Capacité:600 ml
- Seq:06