Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
NucleoMag Kits for Pathogen Detection

NucleoMag Kits for Pathogen Detection

NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 2177,55 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids, Magnetic beads, Plasma, Serum, Cell-free biological fluid, 50 - 100 µl
N° de catalogue MANA744800.4
Fournisseur:   MACHEREY-NAGEL
NucleoMag Kits for Pathogen Detection
N° de catalogue MANA744800.4
Fournisseur:  MACHEREY-NAGEL
Numéro CAS:  
NucleoMag Kits for Pathogen Detection
NucleoMag Kits for Pathogen Detection

NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 2177,55 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids, Magnetic beads, Plasma, Serum, Cell-free biological fluid, 50 - 100 µl

Numéro CAS:  
Prix
1 000,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.

  • Magnetic bead technology
  • NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes

Support protocols for automation platforms available on request.

Handling: Magnetic separation.

Hazardous material: No.

Automation support: Compatible with most open robotic platforms.

Caution: Shelf life: 24 months from date of production. For research use only.

Spécifications

Spécifications

  • Désignation:
    NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 2177,55 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids
  • Format:
    Magnetic beads
  • Volume:
    50 - 100 µl
  • Contenu:
    Plasma, Serum, Cell-free biological fluid
  • Cat. No.:
    MANA744800.4
  • Prep size:
    Scalable
  • Longueur des fragments:
    300 bp - approx. 50 kbp
  • Refine target molecule:
    Viral RNA and DNA
  • Target molecule:
    Copurification of DNA and RNA
  • Method / Format:
    Magnetic beads
  • Operation time:
    40 - 120 min/96 preps
  • Capacité de fixation:
    0,2 µg/µl beads
  • Taille de l'échantillon:
    <200 µl
  • Température de stockage:
    15...25 °C (59...77 °F)

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.