Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
NucleoMag Kits for Pathogen Detection

NucleoMag Kits for Pathogen Detection

NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 601,45 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids, Magnetic beads, Plasma, Serum, Cell-free biological fluid, 50 - 100 µl
N° de catalogue MANA744800.1
Fournisseur:   MACHEREY-NAGEL
NucleoMag Kits for Pathogen Detection
N° de catalogue MANA744800.1
Fournisseur:  MACHEREY-NAGEL
Numéro CAS:  
NucleoMag Kits for Pathogen Detection
NucleoMag Kits for Pathogen Detection

NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 601,45 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids, Magnetic beads, Plasma, Serum, Cell-free biological fluid, 50 - 100 µl

Numéro CAS:  
Prix
268,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.

  • Magnetic bead technology
  • NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes

Support protocols for automation platforms available on request.

Handling: Magnetic separation.

Hazardous material: No.

Automation support: Compatible with most open robotic platforms.

Attention: Shelf life: 24 months from date of production. For research use only.

Spécifications

Spécifications

  • Désignation:
    NucleoMag Virus kit for RNA/DNA purification from cell-free fluids, 601,45 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral nucleic acids
  • Format:
    Magnetic beads
  • Volume:
    50 - 100 µl
  • Contenu:
    Plasma, Serum, Cell-free biological fluid
  • Cat. No.:
    MANA744800.1
  • Prep size:
    Scalable
  • Longueur des fragments:
    300 bp - approx. 50 kbp
  • Refine target molecule:
    Viral RNA and DNA
  • Target molecule:
    Copurification of DNA and RNA
  • Method / Format:
    Magnetic beads
  • Operation time:
    40 - 120 min/96 preps
  • Capacité de fixation:
    0,2 µg/µl beads
  • Taille de l'échantillon:
    <200 µl
  • Température de stockage:
    15...25 °C (59...77 °F)

Spécifications