Dichlorométhane, BAKER ANALYZED® HPLC pour HPLC, J.T.Baker®
Flacon Verre, 2,5 L
Supplier: Avantor
Dichlorométhane, BAKER ANALYZED® HPLC pour HPLC, J.T.Baker®
Catalog # 9410.2500
Supplier: Avantor
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39,8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1,322 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
Documents
Certificates
Search by Lot Numberspec_BAKR9410.1000.pdf spec_BAKR9410.2500.pdf
Product specification
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Specifications
Specification Test Results
- Assay (by GC):min. 99,8 %
- Chloride (Cl):max. 10 ppm
- Density (g/ml) at 25°C:1.315-1.321
- Preservative (Amylene) (ppm):about 20
- Residue after Evaporation (in ppm):max. 2
- Titrable Acid (meq/g):max. 0,0003
- Water (H2O):max. 0,02 %
- Ultraviolet Absorbance (1.00-cm path vs water)::-
- at 254 nm:max. 0,01
- at 280 nm:max. 0,01
- at 350 nm:max. 0,01
- UV Cut-off, nm:max. 233
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



