Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Prednicarbate, Reference Standards

Prednicarbate, Reference Standards

Prednicarbate Related Compound B, USP reference standard, Vial, 20 mg
N° de catalogue 1554920.
Fournisseur:   USP
Prednicarbate, Reference Standards
N° de catalogue 1554920.
Fournisseur:  USP
Numéro CAS:  
Prednicarbate, Reference Standards

Prednicarbate Related Compound B, USP reference standard, Vial, 20 mg

Numéro CAS:  
Prix
509,00 €
Connexion Pour la disponibilité
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    20 mg
  • Normes:
    USP reference standard
  • Désignation:
    Prednicarbate Related Compound B
  • Cat. No.:
    1554920.
  • N ° CAS.:
    104286-02-4
  • Formula:
    C₂₄H₃₂O₇
  • Désignation:
    Prednicarbate Related Compound B
  • Synonyme(s):
    Prednisolone-17-ethylcarbonate

Spécifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.