Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Acetaminophen Related Compounds, Reference Standard

Acetaminophen Related Compounds, Reference Standard

Acetaminophen Related Compound A, USP reference standard, 15 mg
N° de catalogue 1003010.
Fournisseur:   USP
Acetaminophen Related Compounds, Reference Standard
N° de catalogue 1003010.
Fournisseur:  USP
Numéro CAS:  
Acetaminophen Related Compounds, Reference Standard

Acetaminophen Related Compound A, USP reference standard, 15 mg

Numéro CAS:  
Prix
1 150,00 €
Connexion Pour la disponibilité
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Cdt:
    15 mg
  • Normes:
    USP reference standard
  • Désignation:
    Acetaminophen Related Compound A
  • Cat. No.:
    1003010.
  • N ° CAS.:
    2623-33-8
  • Formula:
    C₁₀H₁₁NO₃
  • Désignation:
    Acetaminophen Related Compound A
  • Synonyme(s):
    4-(acetylamino)phenyl acetate

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.