Sample Containers
Sample Containers
-, 82×108×5 mm
N° de catalogue BRBO0244
Fournisseur:   BRABO VERPAKKINGEN
Sample Containers
N° de catalogue BRBO0244
Fournisseur:  BRABO VERPAKKINGEN
Sample Containers
Sample Containers
-, 82×108×5 mm
Prix
49,10 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
PS, clear.
  • Inexpensive, single-use
  • Optimum content visibility
  • Optional matching lids available
Universal container for a wide variety of uses. Ideal for collecting, storing and transporting samples.
Spécifications
  • Capacité:
    -
  • l×p×h:
    82×108×5 mm