Inserts d'injection
Liner, ultra inert, universal, low pressure drop, glass wool, 4 mm
N° de catalogue HEWL5190-3173
Fournisseur: Agilent
Aller à :
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Spécifications
- Ø int.:4 mm
- Désignation:Liner, ultra inert, universal, low pressure drop, glass wool
- Seq:29