Accessories
pour conteneurs 50 l
Catalog # BOCH8344R
Supplier:   BOCHEM
Accessories
Catalog # BOCH8344R
Supplier:  BOCHEM
Accessories
Accessories
pour conteneurs 50 l
Price
293,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Castors for transport container, made of 18/10 stainless steel, with 4 rubber guided rolls, 1 lockable.
Specifications
  • Désignation:
    pour conteneurs 50 l
  • Seq:
    02