Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Transport trolleys

Transport trolleys

Laboratory cart 4, collapsible, 785×450×850 mm
N° de catalogue BOCH11417
Fournisseur:   BOCHEM
Transport trolleys
N° de catalogue BOCH11417
Fournisseur:  BOCHEM
Transport trolleys
Transport trolleys

Laboratory cart 4, collapsible, 785×450×850 mm

Prix
1 510,00 €
Connexion Pour la disponibilité
Product Details & Documents

These transport trolleys are made of stainless steel with two guide and two fixed rolls.

  • Surface matt finish
  • Robust pushing handle

Load capacity: 300 kg

Spécifications
  • Désignation:
    Laboratory cart 4, collapsible
  • Seq:
    01
  • l×p×h:
    785×450×850 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.