Louches
500 ml, Acier inoxydable 18/10
N° de catalogue BOCH3807
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
These portioning ladles are made of 18/10 stainless steel.
Please note: ml indication reached with filling until rim.
Spécifications
- Capacité:500 ml
- Matière:Acier inoxydable 18/10