Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
DEPC

DEPC

Bottle, 5 g
Catalog # RC-041
Irritant
Supplier:   G-Biosciences
DEPC
Catalog # RC-041
Supplier:  G-Biosciences
CAS Number:  1609-47-8
DEPC

Bottle, 5 g

CAS Number:  1609-47-8
Price
60,70 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • CAS Number:1609-47-8
  • Formula:O(COOC₂H₅)₂
  • MDL Number:MFCD00009106
  • Molecular Weight:162.14 g/mol
  • Merck Index:13,08088
  • Boiling Pt:93 °C (1013 hPa)
  • Density:1,123 g/cm³ (20 °C)
  • Flash Pt:69 °C
  • Storage Temperature:Réfrigérateur
  • EINECS:216-542-8
Specifications

Specifications

  • Type de conditionnement:
    Bottle
  • Cdt:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.