Warning labels for hazardous substances
Hazard labels, BPT LABELS 80 X 50 MM IN B-7610, Polyester, noir/jaune/blanc, 262×371 mm, 1 label/card
BRDY801327
: Brady
These self-adhesive signs are designed to warn people to be careful and to take precautions of potentially hazardous substances.
- Couleur:noir/jaune/blanc
- Désignation:Hazard labels
- Dimensions:262×371 mm
- Matière:Polyester
- Seq:04
- Modèle:BPT LABELS 80 X 50 MM IN B-7610
- Emballage:1 label/card



